Learn More
Description
The SNF2-related family of proteins all contain conserved helicase motifs, and have been implicated in transcription regulation, DNA repair, and DNA recombination. The DNA-dependent ATPase activity of these proteins is usually activated by single-stranded, double-stranded, or nucleosomal DNA. The chromodomain helicase DNA binding proteins (CHDs) are a subfamily of the SNF2 family, and includes CHD1, CHD2, CHD3, and CHD4. CHD3 and CHD4 contain N-terminal paired PHD zinc finger motifs (PZF), two heterochromatin binding domains (chromodomains; CDs), and a central SNF2-like helicase/ATPase domain. CHD3 mRNA is widely expressed, and CHD3 protein localizes to the nucleus, but not nucleoli. Both CHD3 and CHD4 were first identified as self-antigens recognized by sera from patients with the autoimmune connective-tissue disease Mi-2 dermatomyositis. In addition, both CHD3 and CHD4 are associated with a nucleosome remodelling and deacetylating (NRD) complex, and facilitate deacetylation of oligonucleosomal histones in vitro. Thus, CHD3 and CHD4 are chromodomain containing proteins that may have important roles in ATP-dependent nucleosome remodelling.
Host Species: Mouse
Clone: 44
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human CHD3 aa. 1277-1474
Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | CHD3 |
| Applications | Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 44 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human CHD3 aa. 1277-1474 |
| Purification Method | Affinity Purified |
| Quantity | 150 μg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.