Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chd2 Rat anti-Human, Mouse, Clone: 8H3, MilliporeSigma™

Catalog No. MABE873MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE873MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE873MI Supplier MilliporeSigma Supplier No. MABE873
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-Chd2 antibody, clone 8H3 is validated for use in western blotting, immunoprecipitation, immunocytochemistry and Chimmunoprecipitation for the detection of Chd2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD3
Applications ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8H3
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001074814
Gene Symbols Chd2
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Chd2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.