Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHIP Mouse anti-Human, Clone: 2F2.1, MilliporeSigma™

Catalog No. MABS133MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS133MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS133MI Supplier MilliporeSigma Supplier No. MABS133

Mouse Monoclonal Antibody

Anti-CHimmunoprecipitation Antibody, clone 2F2.1 detects level of CHimmunoprecipitation and has been published and validated for use in immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHIP
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2F2.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_005852
Gene Symbols STUB1, CHIP, PP1131
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to mouse CHIP.
Purification Method Purifed by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.