Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CHIP, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC711100UL
Description
Specifically detects CHIP Clone: in Drosophila, Hamster, Human, Monkey, Mouse, Rat samples, and it is validated for Immunofluorescence, Immunoprecipitation
CHIP is a U-box dependent E3 ubiquitin ligase and a co-chaperone protein that interacts with Hsc70 and Hsp90 regulating protein triage decisions between folding and degradation. This occurs through CHIP remodeling the complexes to favor substrate degradation thus providing a molecular link between the chaperone and proteosome machinery. CHIP has been reported to be involved in the degradation of c-ErbB2/c-Neu, the glucocorticoid receptor, and the cystic fibrosis transmembrane-conductance regulator. CHIP expression shortens the half-life of both the nascent and mature ErbB2 protein.
Specifications
CHIP | |
Polyclonal | |
Undiluted serum. | |
a recombinant protein consisting of full-length human CHIP fused to a His·Tag™ sequence | |
RUO | |
Recognizes the ∽35kDa CHIP protein. May also detect bands at ∽43, ∽51, and ∽70kDa corresponding to the mono-ubiquitinated, di-ubiquitinated, and dimeric forms of CHIP, respectively. | |
−20°C | |
IgG |
Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
Drosophila, Hamster, Human, Monkey, Mouse, Rat | |
Undiluted |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction