Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHIP/STUB1 Goat anti-Human, Chimpanzee, Polyclonal, MilliporeSigma™

Catalog No. AB10000MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10000MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10000MI Supplier MilliporeSigma Supplier No. AB10000
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Anti-CHIP/STUB1 Antibody is validated for use in ELISA, western blotting for the detection of CHIP/STUB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHIP/STUB1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Gene Symbols STUB1; SDCCAG7; NY-CO-7; HSPABP2; UBOX1; CHIP
Host Species Goat
Immunogen Peptide with sequence DAFISENGWVEDY, from the C-terminus of the protein sequence according to NP_005852.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005861.2
Target Species Chimpanzee, Human
Content And Storage −20°C
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.