Learn More
Description
Cell cycle events are regulated by the sequential activation and deactivation of cyclin dependent kinases (Cdks) and by proteolysis of cyclins. Chk1 and Chk2 are involved in these processes as regulators of Cdks. Chk1 and Chk2 both functionas essential components in the G2 DNA damage checkpoint by phophorylating Cdc25C in response to DNA damage. Phosphorylation inhibits Cdc25C activity, thereby blocking mitosis. Cdc25A, Cdc25B and Cdc25C protein tyrosine phosphatases function as mitotic activators by dephosphorylating Cdc2 p34 on regulatory tyrosine residues. It has also been shown that Chk1 can phosphorylate Wee1 in vitro, providing evidence that the hyperphosphorylated form of Wee1, seen in cells delayed by Chk1 overexpression, is due to phosphorylation by Chk1.
Host Species: Mouse
Clone: DCS-270
Isotype: IgG2a/κ
Cross Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human Chk2 protein.
Molecular Weight: ∽70kDa
Positive Control: HeLa cells. Tonsil.
Cellular Localization: Nuclear
Immunohistochemistry (Formalin/Paraffin), Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | Chk2 Ab-5 |
| Classification | Monoclonal |
| Concentration | n/a |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human Chk2 protein. |
| Quantity | 7 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.