Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk2 Mouse, Unlabeled, Clone: 19, BD

Catalog No. BDB611570 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611570 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611570 Supplier BD Biosciences Supplier No. 611570
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The cell cycle is regulated by multiple checkpoints that determine cell fate. Such checkpoints ensure that DNA replication and chromosomal segregation are completed with high fidelity. When DNA is damaged, specific kinases and phosphatases, key components of cell cycle checkpoints, function to arrest the cell cycle and provide the necessary time for DNA repair. For example, DNA damage induces arrest of the cell cycle at the G2 checkpoint via Wee1-mediated inhibitory phosphorylation of the kinase Cdc2. Inhibition is relieved by the Cdc25C
phosphatase via Cdc2 dephosphorylation. In turn, the Chk1 and Chk2 protein kinases phosphorylate and inhibit Cdc25C, thus preventing activation of the Cdc2-cyclin B complex and entry into mitosis. Chk2, the human homolog of S. cerevisiae Rad53 and S. pombe Cds1, contains a C-terminal kinase domain, an N-terminal regulatory region that is rich in TQ and SQ pairs, and a forked head-associated domain (FHA) which is found in other cell cycle kinases. Chk2 is expressed during late G1 to M phase and is found in the nucleoplasm. Thus, Chk1
and Chk2 activity may be an important checkpoint that inhibits entry into mitosis in response to DNA damage.

Host Species: Mouse
Clone: 19
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse Chk2 aa. 31-234
Formula Weight [Chemical]: 60kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Chk2
Applications Western Blot
Classification Monoclonal
Clone 19
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Chk2 aa. 31-234
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.