Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chk2 Mouse anti-Human, Mouse, Clone: 7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05649
Description
Anti-Chk2 Antibody, clone 7 is a mouse monoclonal antibody for detection of Chk2 also known as CHK2 checkpoint homolog (S. pombe), serine/threonine-protein kinase CHK2 and has been validated in western blot, immunoprecipitation, immunocytochemistry.
Specifications
Chk2 | |
Monoclonal | |
Unconjugated | |
NP_001005735 | |
Mouse | |
Protein G chromatography | |
Primary | |
Stable for 1 year at from date of receipt. | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
7 | |
Protein G purified mouse IgG1 buffer containing 0.07MTris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide and 30% glycerol. Liquid at −20°C. | |
CHEK2, HuCds1, CHK2, LFS2, RAD53, bA444G7, CDS1, OTTHUMP00000028871, PP1425, Cds1 | |
Mixture of two GST fusion proteins corresponding to residues 1-223 and 217-543 of human Chk2 | |
200 μg | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction