Learn More
Chlamydia trachomatis-EB Goat anti-Human, FITC, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Chlamydia trachomatis-EB |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | IgG fraction conjugated with FITC. Supplied in PBS (0.1 M, pH 7.2) with 10mg/mL BSA as a stabilizer and 0.1% sodium azide. |
| Host Species | Goat |
| Immunogen | L2 and other serovar groups |
| Quantity | 1 Unit |
| Primary or Secondary | Primary |
| Target Species | Human |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.