Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chromatin modifying protein 4A, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017064 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-064 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-064 Supplier Abnova Corporation Supplier No. H00029082B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CHMP4A protein.

CHMP4A belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A (MIM 164010), is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 [PubMed 16730941]).[supplied by OMIM

Sequence: MSGLGRLFGKGKKEKGPTPEEAIQKLKETEKILIKKQEFLEQKIQQELQTAKKYGTKNKRAALQALRRKKRFEQQLAQTDGTLSTLEFQREAIENATTNAEVLRTMELAAQSMKKAYQDMDIDKVDELMTDITEQQEVAQQISDAISRPMGFRDDVDEDELLEELEELEQEELAQELLNVGDKEEEPSVKLPSVPSTHLPAGPAPKVDEDEEALKQLAEWVS

Specifications

Antigen chromatin modifying protein 4A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CHMP4A protein.
Formulation PBS with no preservative; pH 7.4
Gene CHMP4A
Gene Accession No. BC010893
Gene Alias C14orf123/CHMP4/CHMP4B/HSPC134/MGC142093/MGC142095/SNF7/SNF7-1/Shax2
Gene Symbols CHMP4A
Host Species Mouse
Immunogen CHMP4A (AAH10893, 1 a.a. ∼ 222 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29082
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.