Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cholesterol-24 Hydroxylase Mouse anti-Mouse, Human, Rat, Rabbit, Clone: 1A7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2259
Description
Detect Cholesterol-24 Hydroxylase using this Anti-Cholesterol-24 Hydroxylase Antibody, clone1A7 validated for use in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry.
Specifications
Cholesterol-24 Hydroxylase | |
Monoclonal | |
Unconjugated | |
NP_006659 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1A7 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
CH24H, CP46, CYP46, CYP46A1 | |
His-tagged recombinant protein corresponding to human cholesterol-24 hydroxylase. | |
100 μg | |
Human, Mouse, Rabbit, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction