Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Choline Acetyltransferase Goat anti-Guinea Pig, Mouse, Rat, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma AB144
Description
Anti-Choline Acetyltransferase Antibody is an antibody against Choline Acetyltransferase for use in immunohistochemistry and western blotting.
Specifications
Choline Acetyltransferase | |
Polyclonal | |
Goat antisera, liquid, no preservatives. | |
Goat | |
Unpurified | |
Primary | |
Guinea Pig, Mouse, Rat | |
Serum |
Immunohistochemistry, Western Blot | |
Unconjugated | |
CHAT; ChAT; CHOACTase; CMS1A; CMS1A2 | |
Human placental enzyme. | |
50 μL | |
NM_020549.3; NM_020984.2; NM_020985.2; NM_020986.2 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction