Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase Chicken anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB15468MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15468MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15468MI Supplier MilliporeSigma Supplier No. AB15468
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-Choline Acetyltransferase Antibody is an antibody against Choline Acetyltransferase for use in immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS, pH 7.2 with 0.02% sodium azide.
Gene Symbols CHAT; ChAT; CMS1A2; CHOACTase; CMS1A; EC 2.3.1.6
Host Species Chicken
Immunogen Synthetic peptide corresponding to human/mouse ChAT protein.
Purification Method Affinity Purified
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_020549.3; NM_020984.2; NM_020985.2; NM_020986.2
Target Species Mouse
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.