Learn More
Choline Acetyltransferase Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Choline Acetyltransferase |
| Applications | ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized. Contains no preservative. Reconstitute with 20μL of sterile distilled water. Centrifure to remove any insoluble material. |
| Gene Symbols | CHAT; ChAT; CMS1A2; CHOACTase; CMS1A; EC 2.3.1.6 |
| Host Species | Rabbit |
| Immunogen | A 22 amino acid synthetic peptide from porcine ChAT, coupled to KLH. The peptide sequence is GLFSSYRLPGHTQDTLVAQKSS. Due to sequence similarity, the antibody is expected to react with pig, mouse primate and human ChAT. |
| Purification Method | Affinity Purified |
| Quantity | 20 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.