Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase Rabbit anti-Avian, Guinea Pig, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5042MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5042MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5042MI Supplier MilliporeSigma Supplier No. AB5042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Choline Acetyltransferase using this Anti-Choline Acetyltransferase Antibody validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized. Reconstitute with 100μL of sterile distilled water. Contains no preservative.
Gene Symbols CHAT; ChAT; CMS1A2; CHOACTase; CMS1A; EC 2.3.1.6
Host Species Rabbit
Immunogen A 22 amino acid synthetic peptide from porcine ChAT, coupled to KLH. The peptide sequence is GLFSSYRLPGHTQDTLVAQKSS. Due to sequence similarity, the antibody is expected to react with pig, rat, mouse and human ChAT.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_020549.3; NM_020984.2; NM_020985.2; NM_020986.2
Target Species Avian, Guinea Pig, Rat
Content And Storage Lyophilized: −70°C for up to 6 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.