Learn More
Choline Acetyltransferase (ChAT) Rabbit anti-Human, Rat, Mouse, Bat, Feline, Monkey, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Choline Acetyltransferase (ChAT) |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit polyclonal antisera, liquid, no preservatives. |
| Gene Symbols | CHAT; ChAT; CHOACTase; CMS1A; CMS1A2 |
| Host Species | Rabbit |
| Immunogen | Human placental enzyme. |
| Purification Method | Unpurified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.