Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase Mouse anti-Human, Pig, Rat, Clone: 1.B3.9B3, MilliporeSigma™

Catalog No. MAB527050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB527050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB527050UG Supplier MilliporeSigma Supplier No. MAB527050UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Choline Acetyltransferase Antibody, clone 1.B3.9B3 is an antibody against Choline Acetyltransferase for use in ELISA, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1.B3.9B3
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in PBS with 0.1% sodium azide as a preservative.
Gene Accession No. NM_020549.3; NM_020984.2; NM_020985.2; NM_020986.2
Gene Symbols CHAT, ChAT, CMS1A2, CHOACTase, CMS1A, EC 2.3.1.6
Host Species Mouse
Immunogen Highly purified ChAT obtained from porcine brain (Ostermann-Fatif et al., 1990).
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Pig, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.