Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase Mouse anti-Human, Monkey, Rat, Clone: 1E6, MilliporeSigma™

Catalog No. MAB305MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB305MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB305MI Supplier MilliporeSigma Supplier No. MAB305
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Choline Acetyltransferase using this Anti-Choline Acetyltransferase Antibody, clone 1E6 validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase
Applications Immunohistochemistry
Classification Monoclonal
Clone 1E6
Conjugate Unconjugated
Formulation Ascites fluid containing no preservatives.
Gene Accession No. NM_020549.3; NM_020984.2; NM_020985.2; NM_020986.2
Gene Symbols CHAT, ChAT, CMS1A2, CHOACTase, CMS1A, EC 2.3.1.6
Host Species Mouse
Immunogen Choline acetyltransferase purified from rat brain.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Monkey, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.