Learn More
MilliporeSigma™ anti-Choline Acetyltransferase, Clone: 1E6, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB305/DEL
Description
Immunohistochemistry
Specifications
Choline Acetyltransferase | |
Unconjugated | |
Choline acetyltransferase purified from rat brain. | |
RUO | |
Primary | |
Ascites |
Monoclonal | |
Mouse | |
100 μL | |
Neuroscience | |
Human, Monkey, Rat | |
IgG1 |