Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase Mouse anti-Guinea Pig, Human, Monkey, Rat, Clone: 28C4, MilliporeSigma™

Catalog No. MAB5350MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5350MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5350MI Supplier MilliporeSigma Supplier No. MAB5350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Choline Acetyltransferase Antibody, clone 28C4 detects level of Choline Acetyltransferase and has been published and validated for use in ELISA, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 28C4
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid.
Gene Accession No. NM_020549.3; NM_020984.2; NM_020985.2; NM_020986.2
Gene Symbols CHAT, ChAT, CMS1A2, CHOACTase, CMS1A, EC 2.3.1.6
Host Species Mouse
Immunogen Purified human placenta ChAT.
Quantity 100 μg
Primary or Secondary Primary
Target Species Guinea Pig, Human, Monkey, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.