Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase Mouse anti-Human, MilliporeSigma™

Catalog No. MAB5422MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5422MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5422MI Supplier MilliporeSigma Supplier No. MAB5422

Mouse Monoclonal Antibody

Anti-Choline Acetyltransferase Antibody detects level of Choline Acetyltransferase and has been published and validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M Phosphate buffer containing 0.25M NaCl and 0.1% sodium azide.
Gene Accession No. NM_020549.3; NM_020984.2; NM_020985.2; NM_020986.2
Gene Symbols CHAT, ChAT, CMS1A2, CHOACTase, CMS1A, EC 2.3.1.6
Host Species Mouse
Immunogen Recombinant human ChAT.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.