Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chondroitin 6 Sulfate Mouse anti-Bovine, Canine, Human, Mouse, Rat, Rabbit, Sheep, Clone: MK-302, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2035
Description
Anti-Chondroitin 6 Sulfate Antibody, clone MK-302 is an antibody against Chondroitin 6 Sulfate for use in ELISA, RIA and western blotting.
Specifications
Chondroitin 6 Sulfate | |
Monoclonal | |
Unconjugated | |
NM_001135.2; NM_013227.2 | |
Mouse | |
100 μL | |
Bovine, Canine, Human, Mouse, Rabbit, Rat, Sheep | |
Ascites |
ELISA, Radioimmune Assays (RIA), Western Blot | |
MK-302 | |
Ascites liquid; no preservative. | |
ACAN, MSK16, SEDK, CSPCP, AGCAN, AGC1, CSPG1, CSPGCP | |
Chondroitinase ABC-digested adult human aggrecan. | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction