Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin Sulfate Proteoglycan Mouse anti-Human, Clone: 9.2.27, MilliporeSigma™

Catalog No. MAB2029MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2029MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2029MI Supplier MilliporeSigma Supplier No. MAB2029
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Chondroitin Sulfate Proteoglycan using this Anti-Chondroitin Sulfate Proteoglycan Antibody, clone 9.2.27 validated for use in flow cytometry, immunofluorescence and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondroitin Sulfate Proteoglycan
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 9.2.27
Conjugate Unconjugated
Formulation Purified from ascites by Protein A Sepharose™ chromatography. Purified immunoglobulin in 0.02M PBS pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Gene Accession No. NM_001897.4
Gene Symbols CSPG4, MSK16, MCSP, NG2, MCSPG, MEL-CSPG, HMW-MAA
Host Species Mouse
Immunogen Human M14 melanoma cell extract depleted of fibronectin and bound by lens culinaris lectin-Sepharose™ (Morgan et al., 1981).
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.