Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin Sulfate Proteoglycan Mouse anti-Guinea Pig, Human, Hamster, Rat, Feline, Gerbil, Primate, Clone: Cat-301, MilliporeSigma™

Catalog No. MAB5284MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5284MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5284MI Supplier MilliporeSigma Supplier No. MAB5284
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Chondroitin Sulfate Proteoglycan Antibody, Brain (core protein), clone Cat-301 detects level of Chondroitin Sulfate Proteoglycan and has been published and validated for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondroitin Sulfate Proteoglycan
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Cat-301
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid. Buffer = 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.
Gene Accession No. NM_001135.2; NM_013227.2
Gene Symbols ACAN, MSK16, SEDK, CSPCP, AGCAN, AGC1, CSPG1, CSPGCP
Host Species Mouse
Immunogen Feline spinal cord fixed gray matter.
Quantity 100 μg
Primary or Secondary Primary
Target Species Feline, Gerbil, Guinea Pig, Hamster, Human, Primate, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.