Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHP, Mouse, Clone: 3G8, Abnova™

Catalog No. 89123127 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-127 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-127 Supplier Abnova Corporation Supplier No. H00011261M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CHP.

This gene encodes a phosphoprotein that binds to the Na+/H+ exchanger NHE1. This protein serves as an essential cofactor which supports the physiological activity of NHE family members and may play a role in the mitogenic regulation of NHE1. The protein shares similarity with calcineurin B and calmodulin and it is also known to be an endogenous inhibitor of calcineurin activity. [provided by RefSeq]

Sequence: MESHSVTQAGVQWRDLGSLQPLPPGFKQFSHLSLPSSWDYRRVPPYLGNFCIFSGEGVSPCWPGWS

Specifications

Antigen CHP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CHP.
Formulation ascites with no preservative
Gene CHP
Gene Accession No. BC008373
Gene Alias SLC9A1BP
Gene Symbols CHP
Host Species Mouse
Immunogen CHP (AAH08373, 1 a.a. ∼ 66 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11261
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.