Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHPF Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526056
Description
CHPF is a protein that has both beta-1,3-glucuronic acid and beta-1,4-N-acetylgalactosamine transferase activity. Transfers glucuronic acid (GlcUA) from UDP-GlcUA and N-acetylgalactosamine (GalNAc) from UDP-GalNAc to the non-reducing end of the elongating chondroitin polymer.
Specifications
CHPF | |
Polyclonal | |
CHPF | |
CHSY2, CSS2, N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase II, N-acetylgalactosaminyltransferase 2, chondroitin glucuronyltransferase 2, chondroitin sulfate synthase 2, chondroitin-polymerizing factor, glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
74241, 79586 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8IZ52, Q6IQX7 | |
CHPF | |
KLH conjugated synthetic peptide between 327-354 amino acids from the central region of human CHPF | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction