Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chordin-like 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018798 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-798 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-798 Supplier Abnova Corporation Supplier No. H00091851D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CHRDL1 protein.

This gene encodes an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq

Sequence: MGGMKYIFSLLFFLLLEGGKTEQVKHSETYCMFQDKKYRVGERWHPYLEPYGLVYCVNCICSENGNVLCSRVRCPNVHCLSPVHIPHLCCPRCPDSLPPVNNKVTSKSCEYNGTTYQHGELFVAEGLFQNRQPNQCTQCSCSEGNVYCGLKTCPKLTCAFPVSVPDSCCRVCRGDGELSWEHSDGDIFRQPANREARHSYHRSHYDPPPSRQAGGLSRFPGARSHRGALMDSQQASGTIVQIVINNKHKHGQVCVSNGKTYSHGESWHPNLRAFGIVECVLCTCNVTKQECKKIHCPNRYPCKYPQKIDGKCCKVCPGKKAKELPGQSFDNKGYFCGEETMPVYESVFMEDGETTRKIALETERPPQVEVHVWTIRKGILQHFHIEKISKRMFEELPHFKLVTRTTLSQWKIFTEGEAQISQMCSSRVCRTELEDLVKVLYLERSEKGHC

Specifications

Antigen chordin-like 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CHRDL1 protein.
Formulation PBS with no preservative; pH 7.4
Gene CHRDL1
Gene Accession No. ENST00000372045
Gene Alias CHL/NRLN1/VOPT/dA141H5.1
Gene Symbols CHRDL1
Host Species Rabbit
Immunogen CHRDL1 (ENSP00000361115, 1 a.a. ∼ 450 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 91851
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.