Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNA4, Mouse, Clone: 4E3, Abnova™

Catalog No. 89021082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-082 Supplier Abnova Corporation Supplier No. H00001137M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CHRNA4.

This gene encodes a nicotinic acetylcholine receptor, which belongs to a superfamily of ligand-gated ion channels that play a role in fast signal transmission at synapses. These pentameric receptors can bind acetylcholine, which causes an extensive change in conformation that leads to the opening of an ion-conducting channel across the plasma membrane. This protein is an integral membrane receptor subunit that can interact with either nAChR beta-2 or nAChR beta-4 to form a functional receptor. Mutations in this gene cause nocturnal frontal lobe epilepsy type 1. Polymorphisms in this gene that provide protection against nicotine addiction have been described. [provided by RefSeq

Sequence: HVETRAHAEERLLKKLFSGYNKWSRPVANISDVVLVRFGLSIAQLIDVDEKNQMMTTNVWVKQEWHDYKLRWDPADYENVTSIRIPSELIWRPDIVLYNN

Specifications

Antigen CHRNA4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CHRNA4.
Formulation PBS with no preservative; pH 7.4
Gene CHRNA4
Gene Accession No. NM_000744
Gene Alias BFNC/EBN/EBN1/FLJ95812/NACHR/NACHRA4/NACRA4
Gene Symbols CHRNA4
Host Species Mouse
Immunogen CHRNA4 (NP_000735, 29 a.a. ∼ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1137
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.