Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNB1, Mouse, Clone: 4G4, Abnova™

Catalog No. 89021083 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-083 Supplier Abnova Corporation Supplier No. H00001140M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CHRNB1.

The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome. [provided by RefSeq

Sequence: SEAEGRLREKLFSGYDSSVRPAREVGDRVRVSVGLILAQLISLNEKDEEMSTKVYLDLEWTDYRLSWDPAEHDGIDSLRITAESVWLPDVVLLNNNDGNFDVALD

Specifications

Antigen CHRNB1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CHRNB1.
Formulation PBS with no preservative; pH 7.4
Gene CHRNB1
Gene Accession No. NM_000747
Gene Alias ACHRB/CHRNB/CMS1D/CMS2A/SCCMS
Gene Symbols CHRNB1
Host Species Mouse
Immunogen CHRNB1 (NP_000738, 24 a.a. ∼ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1140
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.