Learn More
Chromogranin A Mouse anti-Human, PE, Clone: S21-537, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564563
Description
Chromogranin A (CGA) is a member of the granin family of regulated secretory proteins that are found in secretory granules in endocrine and neuroendocrine cells and released in response to extracellular stimulation. Intracellularly, granins are important for targeting peptide hormones and neurotransmitters by their ability to aggregate in the low pH, high calcium environment of the trans-Golgi network. Extracellularly, peptides formed from proteolytic processing of granins regulate hormone secretion. CGA is a prohormone that can be cleaved into several biologically active peptides, such as pancreastatin, β-granin, vasostatin, catestatin, and parastatin. β- granin is an N-terminal fragment of CGA, while pancreastatin and catestatin are processed from the central region of CGA. Cells of the adrenal medulla, anterior pituitary, cerebral cortex as well as beta cells of the pancreas and a variety of tumor cell lines express CGA. The expression of CGA can be used to monitor the pancreatic differentiation of pluripotent stem cells.

Specifications
Chromogranin A | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Chromogranin A Recombinant Protein | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
S21-537 | |
CGA; CgA; CHGA; Chromogranin-A; CMGA; SP-I | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.