Learn More
CHST11, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CHST11 peptide using ARM Technology.
Supplier: Abnova Corporation H00050515K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Chondroitin 4-sulfotransferases, such as CHST11, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate to position 4 of N-acetylgalactosamine residues in chondroitin (Yamauchi et al., 2000 [PubMed 10722746]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| CHST11 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| CHST11 | |
| CHST11 | |
| A synthetic peptide of human CHST11 | |
| RUO | |
| 50515 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| carbohydrate (chondroitin 4) sulfotransferase 11 | |
| CHST11 | |
| C4ST, C4ST-1, C4ST1, HSA269537 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.