Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 8, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008081 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-081 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-081 Supplier Abnova Corporation Supplier No. H00064377B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CHST8 protein.

Sulfate groups in carbohydrates confer highly specific functions on glycoproteins, glycolipids, and proteoglycans and are critical for cell-cell interaction, signal transduction, and embryonic development. Sulfotransferases, such as CHST8, carry out sulfation of carbohydrates (Hiraoka et al., 2001 [PubMed 11445554]).[supplied by OMIM

Sequence: MTLRPGTMRLACMFSSILLFGAAGLLLFISLQDPTELAPQQVPGIKFNIRPRQPHHDLPPGGSQDGDLKEPTERVTRDLSSGAPRGRNLPAPDQPQPPLQRGTRLRLRQRRRRLLIKKMPAAATIPANSSDAPFIRPGPGTLDGRWVSLHRSQQERKRVMQEACAKYRASSSRRAVTPRHVSRIFVEDRHRVLYCEVPKAGCSNWKRVLMVLAGLASSTADIQHNTVHYGSALKRLDTFDRQGILHRLSTYTKMLFVREPFERLVSAFRDKFEHPNSYYHPVFGKAILARYRANASREALRTGSGVRFPEFVQYLLDVHRPVGMDIHWDHVSRLCSPCLIDYDFVGKFESMEDDANFFLSLIRAPRNLTFPRFKDRHSQEARTTARIAHQYFAQLSALQRQRTYDFYYMDYLMFNYSKPFADLY

Specifications

Antigen carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CHST8 protein.
Formulation No additive
Gene CHST8
Gene Accession No. NM_022467.3
Gene Alias GALNAC4ST1/GalNAc4ST
Gene Symbols CHST8
Host Species Mouse
Immunogen CHST8 (NP_071912.2, 1 a.a. ∼ 424 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64377
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.