Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chx10 Sheep anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9014MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9014MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9014MI Supplier MilliporeSigma Supplier No. AB9014
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

This Anti-Chx10 Antibody, C-terminus is validated for use in western blotting for the detection of Chx10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chx10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.08% sodium azide.
Gene Symbols VSX2; HOX10; MCOPCB3; CHX10; MCOP2; RET1
Host Species Sheep
Immunogen Recombinant human Chx10 protein, C-terminal.
Quantity 250 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_182894.2
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.