Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Chx10, Polyclonal

Catalog No. AB9016MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB9016MI 250 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9016MI Supplier MilliporeSigma™ Supplier No. AB9016
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Specifically detects Chx10 Clone: in Bovine, Human, Mouse, Rat samples, and it is validated for Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Chx10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.08% sodium azide.
Gene Accession No. P58304
Gene Symbols VSX2, HOX10, MCOPCB3, CHX10, MCOP2, RET1
Host Species Sheep
Immunogen Recombinant human Chx10 protein, N-terminal.
Quantity 250 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NM_182894.2
Test Specificity Chx10
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to six months after date of receipt; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.