Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chymase Mouse anti-Human, Biotin, Clone: B7, MilliporeSigma™

Catalog No. MAB1254BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1254BMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1254BMI Supplier MilliporeSigma Supplier No. MAB1254B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Chymase Antibody, clone B7, biotin conjugated is validated for use in DB, immunohistochemistry for the detection of Chymase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chymase
Applications Dot Blot, Immunohistochemistry
Classification Monoclonal
Clone B7
Conjugate Biotin
Formulation Culture supernatant, purified by ammonium sulfate precipitation and DEAE-Sephacyl chromatography, labeled with biotin. In 0.01M PBS, 15mg/mL BSA, 0.1% sodium azide, pH 7.1.
Gene Accession No. NM_001836.2
Gene Symbols CMA1, CYM, MGC119890, Alpha-chymase, MCT1, MGC119891, CYH, EC 3.4.21.39
Host Species Mouse
Immunogen Chymase purified from human skin.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.