Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ cIAP-2 Rabbit anti-Human, Mouse, Rat, Polyclonal

Catalog No. AB3615MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3615MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3615MI Supplier MilliporeSigma™ Supplier No. AB3615

Rabbit Polyclonal Antibody

Anti-cIAP-2 Antibody detects level of cIAP-2 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cIAP-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS, pH 7.2, containing 50% glycerol, 1% BSA and 0.02% sodium azide.
Gene Symbols BIRC3; RNF49; MALT2; MIHC; HAIP1; hiap-1; C-IAP2; API2; HIAP1; HIAP-1; AIP1; IAP1; CIAP2; cIAP2
Host Species Rabbit
Immunogen Synthetic peptide toward the C - terminal end of cIAP-2
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001165.3; NM_182962.1
Target Species Human, Mouse, Rat
Content And Storage −20°, −70° in ubndiluted aliquotes for long term.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.