Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cell death-inducing DFFA-like effector a, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017520 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-520 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-520 Supplier Abnova Corporation Supplier No. H00001149D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CIDEA protein.

This gene encodes the homolog of the mouse protein Cidea that has been shown to activate apoptosis. This activation of apoptosis is inhibited by the DNA fragmentation factor DFF45 but not by caspase inhibitors. Mice that lack functional Cidea have higher metabolic rates, higher lipolysis in brown adipose tissue and higher core body temperatures when subjected to cold. These mice are also resistant to diet-induced obesity and diabetes. This suggests that in mice this gene product plays a role in thermogenesis and lipolysis. Two alternative transcripts encoding different isoforms have been identified. [provided by RefSeq

Sequence: MRGDRASGGPGNHNGSWAREGPRLGPSWKRGLWSPRGGPNRPAEPSRPLTFMGSQTKRVLFTPLMHPARPFRVSNHDRSSRRGVMASSLQELISKTLDALVIATGLVTLVLEEDGTVVDTEEFFQTLGDNTHFMILEKGQKWMPGSQHVPTCSPPKRSGIARVTFDLYRLNPKDFIGCLNVKATMYEMYSVSYDIRCTGLKGLLRSLLRFLSYSAQVTGQFLIYLGTYMLRVLDDKEERPSLRSQAKGRFTCG

Specifications

Antigen cell death-inducing DFFA-like effector a
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CIDEA protein.
Formulation PBS with no preservative; pH 7.4
Gene CIDEA
Gene Accession No. NM_198289
Gene Alias CIDE-A
Gene Symbols CIDEA
Host Species Rabbit
Immunogen CIDEA (NP_938031.1, 1 a.a. ∼ 253 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 1149
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.