Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ciliary Neurotrophic Factor Mouse anti-Human, Mouse, Monkey, Rat, Clone: 4-68, MilliporeSigma™

Catalog No. MAB338MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB338MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB338MI Supplier MilliporeSigma Supplier No. MAB338
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Ciliary Neurotrophic Factor Antibody, clone 4-68 is validated for use in western blot, immunohistochemistry for the detection of Ciliary Neurotrophic Factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ciliary Neurotrophic Factor
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4-68
Conjugate Unconjugated
Formulation Liquid in 0.02M phosphate buffer, 0.25M NaCl with 0.1% sodium azide, pH 7.6.
Gene Accession No. NM_000614.2
Gene Symbols CNTF, HCNTF
Host Species Mouse
Immunogen Purified ciliary neurotrophic factor from rat sciatic nerve.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Monkey, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.