Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cip1 Mouse anti-Human, Unlabeled, Clone: 70, BD

Catalog No. BDB610233 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610233 50 μg
BDB610234 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610233 Supplier BD Biosciences Supplier No. 610233
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cip1 (p21) was identified as an inhibitor of cdk activity in a quaternary complex that also included Cyclin D, Cdk4, and PCNA. It has subsequently been shown that Cip1 can directly bind to and inhibit each member of the cdk family, though the affinities vary for each enzyme. Several studies indicate that Cip1 expression is regulated by the p53 tumor suppressor protein. For example, following DNA damage, Cip1 becomes transcriptionally induced in a p53-dependent manner. Thus, Cip1 protein may have a prominent role in mediating cell cycle arrest. Cip1 is also a component of active cyclin/cdk kinases. It has been suggested that Cip1-containing enzymes may transition between active and inactive states through changes in Cip1 stoichiometry. Active complexes appear to contain a single Cip1 molecule, while the inactive complexes have multiple Cip1 subunits. When multiple subunits are complexed with a cdk, cyclin, and PCNA, these Cip1 molecules can block the access of cdk-activating kinase (CAK) to cdk, thus preventing its phosphorylation and activation. However, inhibition of cdk activity by Cip1 does not appear to be dependent upon this mechanism. Other studies on DNA replication indicate that Cip1 can inhibit this process in vitro by directly binding to PCNA, a DNA polymerase-δ processivity factor.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cip1
Applications Western Blot
Classification Monoclonal
Clone 70
Concentration 250μg/mL
Conjugate Unconjugated
Description WAF1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.