Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CITED4 Monoclonal Antibody (HT13-2D6.3)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA116984
Description
Suggested positive control: ECV304 cell lysate, transfected whole cell Hep3B lysate.
Cited4 is a member of The CITED family proteins that bind to CBP/p300 transcriptional integrators through their conserved C-terminal acidic domain and function as coactivators. It also interacts with isoforms of the TFAP2 transcription factor, coactivating TFAP2-dependent transcription.
Specifications
CITED4 | |
Monoclonal | |
1 mg/mL | |
TBS with 0.05% sodium azide | |
Q96RK1 | |
CITED4 | |
N-terminal fusion protein to full-length human CITED4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
HT13-2D6.3 | |
Unconjugated | |
CITED4 | |
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4; cbp/p300-interacting transactivator 4; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 4; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4; cited4; cited4 {ECO:0000312; Mrg2; MRG-2; MSG1-related protein 2; RGD:620113}; transcriptional co-activator 4 | |
Mouse | |
Protein G | |
RUO | |
163732 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction