Learn More
Citrulline Rabbit anti-Human, Mouse, Vertebrates, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Citrulline |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 25mM Tris/Tris-HCl, pH 7.4, 137mM NaCl,2.7mM KCl, 0.02% sodium azide before the addition of glycerol to 30% |
| Host Species | Rabbit |
| Immunogen | Linear peptide containing Citrulline. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.