Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrulline Rabbit anti-Mammals, Polyclonal, MilliporeSigma™

Catalog No. AB5612MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5612MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5612MI Supplier MilliporeSigma Supplier No. AB5612

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Anti-Citrulline Antibody is a Rabbit Polyclonal Antibody for the detection of Citrulline and has been validated in ELISA and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Citrulline
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum. Liquid with 0.05% sodium azide
Host Species Rabbit
Immunogen Citrulline-Glutaraldehyde-BSA
Quantity 100 μL
Primary or Secondary Primary
Target Species Mammalia
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.