Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKS1B, Mouse, Clone: 3G8, Abnova™

Catalog No. 89021098 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-098 Supplier Abnova Corporation Supplier No. H00001163M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CKS1B.

CKS1B protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS1B mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects a specialized role for the encoded protein. At least two transcript variants have been identified for this gene, and it appears that only one of them encodes a protein. [provided by RefSeq

Sequence: MSHKQIYYSDKYDDEEFEYRHVMLPKDIAKLVPKTHLMSESEWRNLGVQQSQGWVHYMIHEPEPHILLFRRPLPKKPKK

Specifications

Antigen CKS1B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CKS1B.
Formulation PBS with no preservative; pH 7.4
Gene CKS1B
Gene Accession No. NM_001826
Gene Alias CKS1/PNAS-16/PNAS-18/ckshs1
Gene Symbols CKS1B
Host Species Mouse
Immunogen CKS1B (NP_001817, 1 a.a. ∼ 79 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1163
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.