Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC28 protein kinase regulatory subunit 2, Mouse, Clone: 2G12-2A5, Abnova™

Catalog No. 89000577 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-577 Supplier Abnova Corporation Supplier No. H00001164M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CKS2.

CKS2 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS2 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects specialized role for the encoded protein. [provided by RefSeq

Sequence: MAHKQIYYSDKYFDEHYEYRHVMLPRELSKQVPKTHLMSEEEWRRLGVQQSLGWVHYMIHEPEPHILLFRRPLPKDQQK

Specifications

Antigen CDC28 protein kinase regulatory subunit 2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G12-2A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CKS2.
Formulation PBS with no preservative; pH 7.4
Gene CKS2
Gene Accession No. BC006458
Gene Alias CKSHS2
Gene Symbols CKS2
Host Species Mouse
Immunogen CKS2 (AAH06458, 1 a.a. ∼ 79 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 1164
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.