Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLA-1 Mouse anti-Human, Unlabeled, Clone: 25, BD

Catalog No. BDB610882 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610882 50 μg
BDB610883 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610882 Supplier BD Biosciences Supplier No. 610882
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLA-1 (CD36 and LIMPII Analogous-1) is member of a novel gene family that includes CD36, LIMPII, and SR-BI. CD36 is a cell surface glycoprotein that binds to collagen type I and thrombospondin. LIMPII (lysosomal integral membrane protein II), as its name suggests, is expressed on the membrane of lysosomes. SR-BI (scavenger receptor type B class I) is involved in the selective uptake of cholesterol esters. These proteins include two membrane-anchoring regions, two short cytoplasmic tails, and a large extracellular/luminal domain. CLA-1 mRNA is detected in a wide range of tissues including adrenal glands, liver, and testis. Its expression and similarity with other family members suggest it may play a role in HDL metabolism. However, identification of the CLA-1 receptor on monocytes indicates additional CLA-1 functions in leukocytes.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CLA-1
Applications Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Description CD36 and LIMPII Analogous-1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CLA-1 aa. 104-294
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.