Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Human CLA; Clone: HECA-452; PE, BD

Catalog No. BDB563962 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563962 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563962 Supplier BD Biosciences Supplier No. 563962
Add to Cart
Add to Cart

Host Species: Rat
Clone: HECA-452
Isotype: IgM, κ
Species Reactivity: Hu
Immunogen: Lymphocyte-depleted Stromal Preparation of Tonsil

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CLA
Classification Monoclonal
Clone HECA-452
Conjugate PE
Host Species Rat
Immunogen Lymphocyte-depleted Stromal Preparation of Tonsil
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.