Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Clara Cell Secretory Protein Rabbit anti-Mouse, Rat, Polyclonal

Catalog No. 07623MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-623-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-623-MI Supplier MilliporeSigma™ Supplier No. 07623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Clara Cell Secretory Protein using this Anti-Clara Cell Secretory Protein Antibody validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clara Cell Secretory Protein
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum containing 0.05% sodium azide and 30% glycerol.
Gene Symbols SCGB1A1; UP1; uteroglobin; CC10; CC16; CCPBP; UGB; CCSP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 45-68 of mouse CCSP coupled to KLH
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_013051
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.