Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Mouse anti-Amphibian, Bovine, Human, Pig, Rat, Clone: CHC5.9, MilliporeSigma™

Catalog No. CBL188MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL188MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL188MI Supplier MilliporeSigma Supplier No. CBL188
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Clathrin Antibody, clone CHC5.9 is an antibody against Clathrin for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone CHC5.9
Conjugate Unconjugated
Formulation The antibody was purified by gel filtration and is presented as 100μg of lyophilysed material. Reconstitute with 1ml distilled water. Final buffer composition is PBS, pH 7.4.
Gene Accession No. NM_004859.3
Gene Symbols CLTC, CHC17, CLTCL2, Hc, CLH17, CLH-17, KIAA0034
Host Species Mouse
Immunogen Coated vesicles (clathrin) of bovine brain.
Quantity 100 μg
Primary or Secondary Primary
Target Species Amphibian, Bovine, Human, Pig, Rat
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.