Learn More
Claudin 5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137472
Description
Claudin proteins are a family of proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.
This antibody is predicted to react with bovine, chicken, canine, amphibian, mouse, porcine and rat based on sequence homology.Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is lung tissue.
Specifications
Claudin 5 | |
Polyclonal | |
CLDN5 | |
AWAL, BEC1, CPETRL1, TMVCF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7122 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
O00501 | |
CLDN5 | |
Synthetic peptide derived from C-terminus of mouse claudin-5 | |
7 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.