Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CLCA2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP233482
Description
CLCA2 Polyclonal specifically detects CLCA2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CLCA2 | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q9UQC9 | |
CLCA2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: EELTLSWTAPGEDFDQGQATSYEIRMSKSLQNIQDDFNNAILVNTSKRNPQQAGIREIFTFSPQISTNGPEHQPNGETHESHRIYVAIRAMDRNSLQSAVSNIAQAPLFIPPNSDPVPARDYLILKGVLTAMGL | |
0.1 mL | |
Breast Cancer | |
9635 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CACC, CaCC-3, CACC3, Calcium-activated chloride channel family member 2, Calcium-activated chloride channel protein 3, calcium-activated chloride channel regulator 2, calcium-activated chloride channel-2, chloride channel accessory 2, chloride channel regulator 2, chloride channel, calcium activated, family member 2, CLCA family member 2, chloride channel regulator, CLCRG2, FLJ97885, hCaCC-3, hCLCA2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction